Assay Details
| Gene Symbol: NRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006717526.1 | XP_006717589.1 | 13 | 2516 | 90 | |
| XM_011519755.2 | XP_011518057.1 | 13 | 2495 | 90 | ||
| XM_011519756.1 | XP_011518058.1 | 12 | 2411 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin subunit beta 1 | ITGB1 |
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB |
-236.0kb | Sense | Chr. 10 - 32900318 - 32958365 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105376488 | LOC105376488 | 140.2kb | Anti-sense | Chr. 10 - 33334437 - 33385325 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NRP1
Genomic Map