Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZNF337 antisense RNA 1 | ZNF337-AS1 | -10.3kb | Anti-sense | Chr. 20 - 25624045 - 25678074 on NCBI Build GRCh38 | |
| Downstream | vomeronasal 1 receptor 108 pseudogene | VN1R108P | 43.1kb | Anti-sense | Chr. 20 - 25734229 - 25735114 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map