Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 81 | CCDC81 | -11.8kb | Anti-sense | Chr. 11 - 86374736 - 86423109 on NCBI Build GRCh38 | |
| Downstream | malic enzyme 3 | ME3 | NADP-ME | 3.8kb | Sense | Chr. 11 - 86441108 - 86674745 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map