Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 47 | LRRC47 | -43.7kb | Sense | Chr. 1 - 3780220 - 3796504 on NCBI Build GRCh38 | |
| Downstream | DNA fragmentation factor subunit beta | DFFB |
CAD, CPAN, DFF-40, DFF2, DFF40 |
14.4kb | Anti-sense | Chr. 1 - 3857143 - 3885429 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map