Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 1 interacting protein 1 like | MAPK1IP1L |
C14orf32, MISS, c14_5346 |
-62.8kb | Sense | Chr. 14 - 55051608 - 55070194 on NCBI Build GRCh38 |
| Downstream | DLG associated protein 5 | DLGAP5 |
DLG7, HURP |
13.0kb | Anti-sense | Chr. 14 - 55148116 - 55191678 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map