Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SET domain containing 9 | SETD9 | C5orf35 | Anti-sense | Chr. 5 - 56909260 - 56928900 on NCBI Build GRCh38 | |
| Downstream | MIER family member 3 | MIER3 | 9.7kb | Sense | Chr. 5 - 56919602 - 56952130 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map