Assay Details
| Gene Symbol: LOC105378702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093492.1 | - | 1 - 2 | 157 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TraB domain containing 2B | TRABD2B | TIKI2 | Anti-sense | Chr. 1 - 47760528 - 47997367 on NCBI Build GRCh38 | |
| Downstream | Skint1 like (pseudogene) | SKINT1L |
BTN12, SKINT1, SKINTL, SKINTP |
281.5kb | Anti-sense | Chr. 1 - 48101715 - 48182428 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105378702
Genomic Map