Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CDC42 small effector 2 | CDC42SE2 | SPEC2 | -128.6kb | Anti-sense | Chr. 5 - 131264009 - 131394690 on NCBI Build GRCh38 |
| Downstream | folliculin interacting protein 1 | FNIP1 | 116.5kb | Sense | Chr. 5 - 131641714 - 131797063 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map