Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 181 | CCDC181 | C1orf114 | -17.9kb | Sense | Chr. 1 - 169394870 - 169462221 on NCBI Build GRCh38 |
| Downstream | coagulation factor V | F5 |
FVL, PCCF, RPRGL1, THPH2 |
31.0kb | Sense | Chr. 1 - 169511954 - 169586632 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map