Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oligodendrocytic myelin paranodal and inner loop protein | OPALIN |
HTMP10, TMEM10, TMP10 |
-15.2kb | Anti-sense | Chr. 10 - 96343216 - 96359365 on NCBI Build GRCh38 |
| Downstream | transmembrane 9 superfamily member 3 | TM9SF3 |
EP70-P-iso, SMBP |
141.4kb | Anti-sense | Chr. 10 - 96518110 - 96587052 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map