Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gonadotropin releasing hormone receptor | GNRHR |
GNRHR1, GRHR, HH7, LHRHR, LRHR |
-49.3kb | Anti-sense | Chr. 4 - 67737375 - 67756086 on NCBI Build GRCh38 |
| Downstream |
transmembrane protease, serine 11D |
TMPRSS11D | HAT | 13.3kb | Anti-sense | Chr. 4 - 67820876 - 67885063 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map