Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 3 | TAF3 |
TAF140, TAFII-140, TAFII140 |
Sense | Chr. 10 - 7818477 - 8016631 on NCBI Build GRCh38 | |
| Downstream | GATA binding protein 3 | GATA3 |
HDR, HDRS |
189.5kb | Sense | Chr. 10 - 8045421 - 8075201 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map