Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CSTF3 antisense RNA 1 (head to head) | CSTF3-AS1 | Sense | Chr. 11 - 33161657 - 33191598 on NCBI Build GRCh38 | ||
| Downstream | homeodomain interacting protein kinase 3 | HIPK3 |
DYRK6, FIST3, PKY, YAK1 |
85.6kb | Sense | Chr. 11 - 33256672 - 33357023 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map