Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PLCG1 antisense RNA 1 | PLCG1-AS1 | TOP1-AS1 | Sense | Chr. 20 - 41097994 - 41138000 on NCBI Build GRCh38 | |
| Downstream | phospholipase C gamma 1 | PLCG1 |
NCKAP3, PLC-II, PLC1, PLC148, PLCgamma1 |
37.0kb | Anti-sense | Chr. 20 - 41137519 - 41175721 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map