Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ITFG1 antisense RNA 1 | ITFG1-AS1 | Anti-sense | Chr. 16 - 47144068 - 47162747 on NCBI Build GRCh38 | ||
| Downstream | phosphorylase kinase regulatory subunit beta | PHKB | 304.0kb | Anti-sense | Chr. 16 - 47461131 - 47701523 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map