Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 4159 | 64 | |
| XM_005271139.3 | XP_005271196.1 | 5 | 4107 | 64 | ||
| GenBank mRNA | AK094134.1 | - | 1 | 1445 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | ZFP69 zinc finger protein | ZFP69 |
ZFP69A, ZKSCAN23A, ZNF642, ZSCAN54A |
10.5kb | Sense | Chr. 1 - 40477220 - 40496343 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZFP69B
Genomic Map