Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 35 member E1 | SLC35E1 | -242.0 | Anti-sense | Chr. 19 - 16549837 - 16572382 on NCBI Build GRCh38 | |
| Downstream | small integral membrane protein 7 | SMIM7 | C19orf42 | 55.4kb | Anti-sense | Chr. 19 - 16630741 - 16660177 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map