Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 710 | LINC00710 | -95.5kb | Anti-sense | Chr. 10 - 10934940 - 10952163 on NCBI Build GRCh38 | |
| Downstream | CELF2 antisense RNA 2 | CELF2-AS2 | 22.0kb | Anti-sense | Chr. 10 - 11071541 - 11105488 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map