Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105374378 | LOC105374378 | -15.9kb | Sense | Chr. 2 - 27716145 - 27761935 on NCBI Build GRCh38 | |
| Downstream | ribokinase | RBKS |
RBSK, RK |
1.6kb | Anti-sense | Chr. 2 - 27781364 - 27890396 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map