Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prune homolog 2 | PRUNE2 |
BMCC1, BNIPXL, C9orf65, KIAA0367 |
Sense | Chr. 9 - 76611376 - 76906220 on NCBI Build GRCh38 | |
| Downstream | forkhead box B2 | FOXB2 | bA159H20.4 | 229.0kb | Anti-sense | Chr. 9 - 77019655 - 77020953 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map