Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAH-box helicase 36 | DHX36 |
DDX36, G4R1, MLEL1, RHAU |
-10.3kb | Sense | Chr. 3 - 154275668 - 154324497 on NCBI Build GRCh38 |
| Downstream | G protein-coupled receptor 149 | GPR149 |
IEDA, PGR10 |
625.0 | Sense | Chr. 3 - 154337672 - 154429715 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map