Assay Details
| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518646.2 | XP_011516948.1 | 18 | 4828 | 98 | |
| GenBank mRNA | AK094719.1 | - | 1 | 2350 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 476 | LINC00476 |
C9orf130, NAG11, NAG12 |
-102.3kb | Anti-sense | Chr. 9 - 95806088 - 95875977 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 92 | LINC00092 |
NCRNA00092, bA346B7.1 |
39.0kb | Anti-sense | Chr. 9 - 96019732 - 96021755 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ERCC6L2
Genomic Map