Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin 42 pseudogene | KRT42P | -25.4kb | Anti-sense | Chr. 17 - 41626327 - 41640199 on NCBI Build GRCh38 | |
| Downstream | eukaryotic translation initiation factor 1 | EIF1 |
A121, EIF-1, EIF1A, ISO1, SUI1 |
20.8kb | Sense | Chr. 17 - 41688875 - 41691646 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map