Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZRANB2 antisense RNA 2 (head to head) | ZRANB2-AS2 | -158.2kb | Anti-sense | Chr. 1 - 71081324 - 71237723 on NCBI Build GRCh38 | |
| Downstream | neuronal growth regulator 1 | NEGR1 |
DMML2433, IGLON4, KILON, Ntra |
4.4kb | Sense | Chr. 1 - 71402942 - 72282850 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map