Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aspartate beta-hydroxylase domain containing 1 | ASPHD1 | Anti-sense | Chr. 16 - 29900392 - 29919860 on NCBI Build GRCh38 | ||
| Downstream | transmembrane protein 219 | TMEM219 | IGFBP-3R | 46.7kb | Anti-sense | Chr. 16 - 29962030 - 29973052 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map