Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuraminidase 4 | NEU4 | Anti-sense | Chr. 2 - 241809065 - 241817413 on NCBI Build GRCh38 | ||
| Downstream | programmed cell death 1 | PDCD1 |
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1 |
37.9kb | Sense | Chr. 2 - 241849881 - 241858908 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map