Assay Details
| Gene Symbol: PAWR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095165.1 | - | 1 | 951 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin 1 | SYT1 |
P65, SVP65, SYT |
-132.9kb | Anti-sense | Chr. 12 - 78863993 - 79452008 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |

PAWR
Genomic Map