Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RPARP antisense RNA 1 | RPARP-AS1 | Sense | Chr. 10 - 102449817 - 102456293 on NCBI Build GRCh38 | ||
| Downstream | major facilitator superfamily domain containing 13A | MFSD13A |
C10orf77, TMEM180, bA18I14.8 |
7.8kb | Sense | Chr. 10 - 102460896 - 102480118 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map