Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | POC1B-GALNT4 readthrough | POC1B-GALNT4 | -17.9kb | Sense | Chr. 12 - 89519412 - 89526262 on NCBI Build GRCh38 | |
| Downstream | ATPase plasma membrane Ca2+ transporting 1 | ATP2B1 |
PMCA1, PMCA1kb |
43.6kb | Sense | Chr. 12 - 89588049 - 89709353 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map