| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008061.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155047.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155047.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155047.hg.1 |
TC1800008061.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800155047.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155047.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
NR_110775.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
TC1800008061.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
ENST00000566225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
ENST00000567609.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155050.hg.1 |
NR_110775.1 |
Refseq |