Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nyctalopin | NYX |
CLRP, CSNB1, CSNB1A, CSNB4, NBM1 |
-77.0kb | Anti-sense | Chr. X - 41447460 - 41476414 on NCBI Build GRCh38 |
| Downstream | G protein-coupled receptor 34 | GPR34 | LYPSR1 | 134.2kb | Anti-sense | Chr. X - 41686543 - 41697277 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map