Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929099 | LOC101929099 | -23.8kb | Anti-sense | Chr. 1 - 117025482 - 117059490 on NCBI Build GRCh38 | |
| Downstream | microRNA 942 | MIR942 |
MIRN942, hsa-mir-942, mir-942 |
9.6kb | Sense | Chr. 1 - 117094643 - 117094728 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map