Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 117 | ZNF117 |
H-plk, HPF9 |
-13.9kb | Sense | Chr. 7 - 64974452 - 64991036 on NCBI Build GRCh38 |
| Downstream | chaperonin containing TCP1 subunit 6 pseudogene 3 | CCT6P3 | CCT6-3P | 31.7kb | Anti-sense | Chr. 7 - 65038354 - 65074713 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map