| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000237654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000504697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000505609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000506614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000507788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000511943.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000513774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
ENST00000515790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011087.hg.1 |
NM_006835.2 |
Refseq |