Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 152 | GPR152 | PGR5 | Anti-sense | Chr. 11 - 67451301 - 67452729 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 134 | TMEM134 | 10.7kb | Anti-sense | Chr. 11 - 67464348 - 67469277 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map