Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
actin, alpha 1, skeletal muscle |
ACTA1 |
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM |
-6.2kb | Sense | Chr. 1 - 229431245 - 229434096 on NCBI Build GRCh38 |
| Downstream | nucleoporin 133 | NUP133 | hNUP133 | 228.0 | Sense | Chr. 1 - 229440995 - 229508343 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map