Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation initiation factor 4E family member 3 | EIF4E3 |
eIF-4E3, eIF4E-3 |
Anti-sense | Chr. 3 - 71679289 - 71754831 on NCBI Build GRCh38 | |
| Downstream | G protein-coupled receptor 27 | GPR27 | SREB1 | 24.4kb | Sense | Chr. 3 - 71754050 - 71755177 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map