Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transient receptor potential cation channel subfamily M member 3 | TRPM3 |
GON-2, LTRPC3, MLSN2 |
Anti-sense | Chr. 9 - 70529063 - 71446989 on NCBI Build GRCh38 | |
| Downstream | microRNA 204 | MIR204 |
MIRN204, RDICC, miRNA204, mir-204 |
201.7kb | Anti-sense | Chr. 9 - 70809975 - 70810084 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map