Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S16 pseudogene 5 | RPS16P5 | RPS16_3_702 | -8.7kb | Sense | Chr. 6 - 53334460 - 53337617 on NCBI Build GRCh38 |
| Downstream | glutamate-cysteine ligase catalytic subunit | GCLC |
GCL, GCS, GLCL, GLCLC |
149.4kb | Sense | Chr. 6 - 53497341 - 53545129 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map