Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 50 | CCDC50 |
C3orf6, DFNA44, YMER |
Sense | Chr. 3 - 191329082 - 191398670 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LincR-0002 | LINCR-0002 | 52.4kb | Sense | Chr. 3 - 191425524 - 191590356 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map