Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 149 | GPR149 |
IEDA, PGR10 |
-658.7kb | Anti-sense | Chr. 3 - 154337672 - 154429715 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1487 | LINC01487 | 150.8kb | Sense | Chr. 3 - 155240945 - 155243124 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map