Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 18 | CCDC18 | NY-SAR-41 | Sense | Chr. 1 - 93179802 - 93279037 on NCBI Build GRCh38 | |
| Downstream | CCDC18 antisense RNA 1 | CCDC18-AS1 | 127.2kb | Anti-sense | Chr. 1 - 93310109 - 93345811 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map