Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC729220 | FLJ45513 | -39.3kb | Sense | Chr. 17 - 49845910 - 49848837 on NCBI Build GRCh38 | |
| Downstream | distal-less homeobox 4 | DLX4 |
BP1, DLX7, DLX8, DLX9, OFC15 |
73.1kb | Sense | Chr. 17 - 49968970 - 49974959 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map