Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8 family member C | LRRC8C |
AD158, FAD158 |
-131.2kb | Sense | Chr. 1 - 89615810 - 89719535 on NCBI Build GRCh38 |
| Downstream | gem nuclear organelle associated protein 8 pseudogene 4 | GEMIN8P4 | 140.3kb | Anti-sense | Chr. 1 - 89993265 - 89994966 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map