Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SIX homeobox 3 | SIX3 | HPE2 | -1.1kb | Sense | Chr. 2 - 44941898 - 44946077 on NCBI Build GRCh38 |
| Downstream | SIX homeobox 2 | SIX2 | 36.4kb | Anti-sense | Chr. 2 - 45005185 - 45009645 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map