Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and SOCS box containing 1 | ASB1 | ASB-1 | Anti-sense | Chr. 2 - 238426741 - 238452250 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1107 | LINC01107 | 83.8kb | Sense | Chr. 2 - 238510690 - 238555499 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map