Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 4 | ST13P4 |
FAM10A4, FAM10A4P |
-204.7kb | Anti-sense | Chr. 13 - 50172018 - 50173616 on NCBI Build GRCh38 |
| Downstream | DLEU1 antisense RNA 1 | DLEU1-AS1 | LINC01308 | 141.6kb | Sense | Chr. 13 - 50520933 - 50527449 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map