Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1210 | LINC01210 | -11.1kb | Sense | Chr. 3 - 137771911 - 137780878 on NCBI Build GRCh38 | |
| Downstream | claudin 18 | CLDN18 |
SFTA5, SFTPJ |
202.1kb | Sense | Chr. 3 - 137998816 - 138033652 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map