Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507557 | LOC100507557 | Anti-sense | Chr. 6 - 145734869 - 145886585 on NCBI Build GRCh38 | ||
| Downstream | F-box protein 30 | FBXO30 | Fbx30 | 2.4kb | Sense | Chr. 6 - 145798135 - 145864409 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map