Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 31 | STK31 |
SGK396, TDRD8 |
-454.1kb | Sense | Chr. 7 - 23710141 - 23832513 on NCBI Build GRCh38 |
| Downstream | membrane palmitoylated protein 6 | MPP6 |
PALS2, VAM-1, VAM1, p55T |
284.7kb | Sense | Chr. 7 - 24573346 - 24694086 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map